Thermo Scientific Chemicals

Magnesium acetate tetrahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 270500050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

Magnesium acetate tetrahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 270500050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 270500050
Price (USD)
Price: 104.00
Online price: 90.65
Your price:
Quantity
-

Chemical Identifiers

CAS
16674-78-5
IUPAC Name
magnesium(2+) diacetate tetrahydrate
Molecular Formula
C4H14MgO8
InChI Key
XKPKPGCRSHFTKM-UHFFFAOYSA-L
SMILES
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Appearance (Form)
Crystalline powder
Heavy metals (as Pb)
=<5 ppm
Chloride (Cl)
=<10 ppm
Sulfate (SO4)
=<50 ppm
Copper (Cu)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text