Cadmium sulfate, 99+%, for analysis
Cadmium sulfate, 99+%, for analysis
Cadmium sulfate, 99+%, for analysis
Thermo Scientific Chemicals

Cadmium sulfate, 99+%, for analysis

CAS: 10124-36-4 | CdO4S | 208.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2705600252.5 kgGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 270560025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10124-36-4
IUPAC Namecadmium(2+) sulfate
Molecular FormulaCdO4S
InChI KeyQCUOBSQYDGUHHT-UHFFFAOYSA-L
SMILES[Cd++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
pH3.5 to 6.0 (5 % at 20°C)
Alkalies+earths (Na,K,Ca)=<0.05 %
Iron (Fe)=<5 ppm
Zinc (Zn)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only