Manganese(II) chloride, 97%
Manganese(II) chloride, 97%
Manganese(II) chloride, 97%
Thermo Scientific Chemicals

Manganese(II) chloride, 97%

CAS: 7773-01-5 | Cl2Mn | 125.84 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
1 kg
2.5 kg
Packaging:
檢視全部
Glass bottle
產品號碼 271410250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS7773-01-5
IUPAC Namemanganese(2+) dichloride
Molecular FormulaCl2Mn
InChI KeyGLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Mn++]
檢視更多
規格Specification Sheet規格表
Iron (Fe)=<200 ppm
Appearance (Form)Flakes or beads
Water=<1 %
Nickel (Ni)=<200 ppm
Appearance (Color)Pink
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only