Manganese(II) chloride, 97%
Manganese(II) chloride, 97%
Manganese(II) chloride, 97%
Thermo Scientific Chemicals

Manganese(II) chloride, 97%

CAS: 7773-01-5 | Cl2Mn | 125.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 271412500
Price (USD)
32.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
32.00
Each
Chemical Identifiers
CAS7773-01-5
IUPAC Namemanganese(2+) dichloride
Molecular FormulaCl2Mn
InChI KeyGLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Mn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<200 ppm
Appearance (Form)Flakes or beads
Water=<1 %
Nickel (Ni)=<200 ppm
Appearance (Color)Pink
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only