5-Bromo-4-chloro-3-indolyl phosphate, disodium salt, hydrate, 98%
5-Bromo-4-chloro-3-indolyl phosphate, disodium salt, hydrate, 98%
5-Bromo-4-chloro-3-indolyl phosphate, disodium salt, hydrate, 98%
5-Bromo-4-chloro-3-indolyl phosphate, disodium salt, hydrate, 98%
Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl phosphate, disodium salt, hydrate, 98%

CAS: 205926-98-3 | C8H4BrClNNa2O4P | 370.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 271462500
Price (EUR)
108,65
Online Exclusive
120,00
Save 11,35 (9%)
Each
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
108,65
Online Exclusive
120,00
Save 11,35 (9%)
Each
Chemical Identifiers
CAS205926-98-3
IUPAC Namedisodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
Molecular FormulaC8H4BrClNNa2O4P
InChI KeyOAZUOCJOEUNDEK-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]P([O-])(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige to light blue
Appearance (Form)Powder
Infrared spectrumConforms
Water5 to 9 % (K.F.)
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only