2-Ethylhexylamine, 99%
2-Ethylhexylamine, 99%
2-Ethylhexylamine, 99%
Thermo Scientific Chemicals

2-Ethylhexylamine, 99%

CAS: 104-75-6 | C8H19N | 129.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
View all
Glass bottle
Catalog number 271661000
Price (USD)
26.65
Special offer
Online exclusive
Ends: 30-Jun-2026
30.90
Save 4.25 (14%)
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
26.65
Special offer
Online exclusive
Ends: 30-Jun-2026
30.90
Save 4.25 (14%)
Each
Chemical Identifiers
CAS104-75-6
IUPAC Name3-(aminomethyl)heptane
Molecular FormulaC8H19N
InChI KeyLTHNHFOGQMKPOV-UHFFFAOYNA-N
SMILESCCCCC(CC)CN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4290 to 1.4320 (20°C, 589 nm)
Water=<0.2 %
GC>=98.5 %
Color scale=<20 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only