Sodium phosphate, monobasic dihydrate, 99+%, for analysis
Sodium phosphate, monobasic dihydrate, 99+%, for analysis
Sodium phosphate, monobasic dihydrate, 99+%, for analysis
Thermo Scientific Chemicals

Sodium phosphate, monobasic dihydrate, 99+%, for analysis

CAS: 13472-35-0 | H2NaO42H2O | 156.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 271750010
Price (USD)
94.10
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
94.10
Each
Chemical Identifiers
CAS13472-35-0
IUPAC Namesodium dihydrate dihydrogen phosphate
Molecular FormulaH6NaO6P
InChI KeyVBJGJHBYWREJQD-UHFFFAOYSA-M
SMILESO.O.[Na+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Calcium (Ca)=<50 ppm
Magnesium (Mg)=<10 ppm
Arsenic (As)=<2 ppm
Appearance (Color)White
Titration with NaOH>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only