Sodium phosphate, monobasic dihydrate, 99+%, for analysis
Sodium phosphate, monobasic dihydrate, 99+%, for analysis
Sodium phosphate, monobasic dihydrate, 99+%, for analysis
Thermo Scientific Chemicals

Sodium phosphate, monobasic dihydrate, 99+%, for analysis

CAS: 13472-35-0 | H2NaO42H2O | 156.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 271750100
Price (USD)
493.00
Each
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
493.00
Each
Chemical Identifiers
CAS13472-35-0
IUPAC Namesodium dihydrate dihydrogen phosphate
Molecular FormulaH6NaO6P
InChI KeyVBJGJHBYWREJQD-UHFFFAOYSA-M
SMILESO.O.[Na+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with NaOH>=99.0 %
Loss on drying=<24 %
Heavy metals (as Pb)=<10 ppm
Nitrogen compounds (as N)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only