Thermo Scientific Chemicals

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 99+%, Thermo Scientific Chemicals

Catalog number: 271811000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 99+%, Thermo Scientific Chemicals

Catalog number: 271811000
100 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
6289-46-9
IUPAC Name
1,4-dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
Molecular Formula
C10H12O6
InChI Key
MHKKFFHWMKEBDW-UHFFFAOYNA-N
SMILES
COC(=O)C1CC(=O)C(CC1=O)C(=O)OC
Appearance (Color)
White to yellow-green
HPLC
>=98.0 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
Melting point
153°C to 157°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text