SoftFit-L™ Pipette Tips, Filtered, Low-Retention, Hinged Rack, PCR Ready
SoftFit-L™ Pipette Tips, Filtered, Low-Retention, Hinged Rack, PCR Ready
SoftFit-L™ Pipette Tips, Filtered, Low-Retention, Hinged Rack, PCR Ready
SoftFit-L™ Pipette Tips, Filtered, Low-Retention, Hinged Rack, PCR Ready
SoftFit-L™ Pipette Tips, Filtered, Low-Retention, Hinged Rack, PCR Ready
SoftFit-L™ Pipette Tips, Filtered, Low-Retention, Hinged Rack, PCR Ready
SoftFit-L™ Pipette Tips, Filtered, Low-Retention, Hinged Rack, PCR Ready

SoftFit-L™ Pipette Tips, Filtered, Low-Retention, Hinged Rack, PCR Ready

Pipette samples with the accuracy and precision you expect from your pipette, every time, with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Unit Size
2739-05-HRPK
also known as 2739-05-HR
300 μLPack of 960
2739-05-HR300 μLCase of 4800
2749-05-HR20 μLCase of 4800
2749-05-HRPK20 μLPack of 960
2769-05-HR200 μLCase of 4800
2769-05-HRPK200 μLPack of 960
2779-05-HR1000 μLCase of 3072
2779-05-HRPK1000 μLPack of 768
2789-05-HR1200 μLCase of 3072
2789-05-HRPK
also known as 2789-05-HR
1200 μLPack of 768
Catalog number 2739-05-HRPK
also known as 2739-05-HR
Price (KRW)
187,000
Pack of 960
Volume (Metric):
300 μL
Unit Size:
Pack of 960
Price (KRW)
187,000
Pack of 960
Compatible with Rainin™ LTS pipettes. Provide outstanding accuracy and precision, every time.
  • Low-retention tips maximize sample delivery
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allows quick identification of tip volume
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description300L, Filtered, Sterile, Hinged Rack
Length (English)2.26 in.
Length (Metric)5.75cm
Model300L
ColorNatural
Color CodeOrange
FilteredFiltered
Fit for PurposeSterile & PCR Ready
For Use With (Equipment)Rainin LTS LiteTouch
FormatHinged Rack
Product LineART SoftFit L
SterilitySterile
Tip StyleUniversal
Volume (Metric)300 μL
Unit SizePack of 960