BOC-L-Proline, 99+%
BOC-L-Proline, 99+%
BOC-L-Proline, 99+%
Thermo Scientific Chemicals

BOC-L-Proline, 99+%

CAS: 15761-39-4 | C10H17NO4 | 215.249 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 275650250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS15761-39-4
IUPAC Name1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid
Molecular FormulaC10H17NO4
InChI KeyZQEBQGAAWMOMAI-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 % (K.F.)
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Appearance (Color)White
Melting point133°C to 136°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only