Nalpha-BOC-L-Tryptophane, 97%
Nalpha-BOC-L-Tryptophane, 97%
Nalpha-BOC-L-Tryptophane, 97%
Acros Organics

Nalpha-BOC-L-Tryptophane, 97%

CAS: 13139-14-5 | C16H20N2O4 | 304.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass Bottle
Catalog number 275690250
also known as 27569
HSN 29224990
Price (INR)
16,878.00
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS13139-14-5
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid
Molecular FormulaC16H20N2O4
InChI KeyNFVNYBJCJGKVQK-ZDUSSCGKSA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<1.0 % (K.F.)
Appearance (Color)White to off-white
Titration with NaOH>=96.0 %
HPLC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only