BOC-L-Tyrosine, 99+%
BOC-L-Tyrosine, 99+%
BOC-L-Tyrosine, 99+%
Thermo Scientific Chemicals

BOC-L-Tyrosine, 99+%

CAS: 3978-80-1 | C14H19NO5 | 281.31 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass Bottle
製品番号(カタログ番号) 275701000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3978-80-1
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC14H19NO5
InChI KeyCNBUSIJNWNXLQQ-LDGXTIHJNA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
さらに表示
HPLC>=99.0 %
Water=<1.0 % (K.F.)
Infrared spectrumConforms
Specific optical rotation+36.5° to +39.5° (24°C, 589 nm) (c=1, dioxane)
Appearance (Form)Powder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only