2,2'-Oxybis(ethylamine), 98%
2,2'-Oxybis(ethylamine), 98%
2,2'-Oxybis(ethylamine), 98%
Thermo Scientific Chemicals

2,2'-Oxybis(ethylamine), 98%

CAS: 2752-17-2 | C4H12N2O | 104.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2758400101 gGlass bottle
Catalog number 275840010
Price (USD)
130.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
130.00
Each
Chemical Identifiers
CAS2752-17-2
IUPAC Name2-(2-aminoethoxy)ethan-1-amine
Molecular FormulaC4H12N2O
InChI KeyGXVUZYLYWKWJIM-UHFFFAOYSA-N
SMILESNCCOCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4550 to 1.4580 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only