2-Methylindoline, 99+%
2-Methylindoline, 99+%
2-Methylindoline, 99+%
2-Methylindoline, 99+%
Thermo Scientific Chemicals

2-Methylindoline, 99+%

CAS: 6872-06-6 | C9H11N | 133.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 275881000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6872-06-6
IUPAC Name2-methyl-2,3-dihydro-1H-indole
Molecular FormulaC9H11N
InChI KeyQRWRJDVVXAXGBT-UHFFFAOYNA-N
SMILESCC1CC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
GC>=99.0 %
Water=<0.5 % (K.F.)
Refractive index1.5680 to 1.5700 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only