Casein, pure
Casein, pure
Casein, pure
Thermo Scientific Chemicals

Casein, pure

Casein, CAS # 9000-71-9, is a family of related phosphoproteins, usually isolated from bovine milk. | CAS: 9000-71-9 | C81H125N22O39P | 2061.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
25 kg
Packaging:
View all
Plastic Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 276070010
Price (EUR)
86,65
Online exclusive
95,60
Save 8,95 (9%)
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (EUR)
86,65
Online exclusive
95,60
Save 8,95 (9%)
Each
Chemical Identifiers
CAS9000-71-9
MDL NumberMFCD00081481
SpecificationsSpecification SheetSpecification Sheet
Protein>=92 % (%N x 6.38) On dry substance
Ash=<3 % (800°C)
Impurity=<2.5 % (Fat)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information
• Casein contains a mixture of related phosphoproteins present in bovine milk and cheese. It contains all of the common amino acids and is rich in the essential ones.

Applications

• Casein is a source of amino acids, carbohydrates, and two inorganic elements namely calcium and phosphorus
• This compound can be used in vivo as a solid food sample and in vitro digestion experiments with rodents
• It can increase the photostability of quantum dots and decrease nonspecific binding in immunofluorescence experiments
• Casein, in biochemical experiments, can be used to prepare P407-casein hydrogels and to study the mechanical effects of the addition of casein to P407

Literature References

Tundisi, L.; Yang, R.; Borelli, L.; Alves, T.; Mehta, M.; Chaud, M.; Mazzola, P.; Kohane, D. Enhancement of the mechanical and drug-releasing properties of poloxamer 407 hydrogels with casein. Pharm Res. 2021, 38 (3), 515–522.

Irshad, I.; Kanekanian, A.; Peters, A.; Masud, T. Antioxidant activity of bioactive peptides derived from bovine casein hydrolysate fractions. J. Food Sci. Technol. 2015, 52 (1), 231-239.

Sharma, A. K.; Sawhney, I. K. Modelling moisture sorption characteristics in dried acid casein using connectionist paradigm vis-à-vis classical methods. J. Food Sci. Technol. 2015, 52 (1), 151-160.

RUO – Research Use Only

General References:

  1. Tundisi, L.; Yang, R.; Borelli, L.; Alves, T.; Mehta, M.; Chaud, M.; Mazzola, P.; Kohane, D. Enhancement of the mechanical and drug-releasing properties of poloxamer 407 hydrogels with casein. Pharm Res. 2021, 38 (3), 515–522.
  2. Irshad, I.; Kanekanian, A.; Peters, A.; Masud, T. Antioxidant activity of bioactive peptides derived from bovine casein hydrolysate fractions. J. Food Sci. Technol. 2015, 52 (1), 231-239.
  3. Sharma, A. K.; Sawhney, I. K. Modelling moisture sorption characteristics in dried acid casein using connectionist paradigm vis-à-vis classical methods. J. Food Sci. Technol. 2015, 52 (1), 151-160.