o-Anisoyl chloride, 97%
o-Anisoyl chloride, 97%
o-Anisoyl chloride, 97%
Thermo Scientific Chemicals

o-Anisoyl chloride, 97%

CAS: 21615-34-9 | C8H7ClO2 | 170.6 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 276110250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS21615-34-9
IUPAC Name2-methoxybenzoyl chloride
Molecular FormulaC8H7ClO2
InChI KeyRZNHSEZOLFEFGB-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.5714 to 1.5734 (20°C, 589 nm)
Appearance (Color)Clear yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only