Thermo Scientific Chemicals

2-Amino-1-benzylbenzimidazole, 99+%, Thermo Scientific Chemicals

Catalog number: 276290010
1 g, Each
Thermo Scientific Chemicals

2-Amino-1-benzylbenzimidazole, 99+%, Thermo Scientific Chemicals

Catalog number: 276290010
1 g, Each
Quantity

Chemical Identifiers

CAS
43182-10-1
IUPAC Name
1-benzyl-1H-1,3-benzodiazol-2-amine
Molecular Formula
C14H13N3
InChI Key
LXIDSOCBAAMGJX-UHFFFAOYSA-N
SMILES
NC1=NC2=CC=CC=C2N1CC1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Titration with HClO4
>=99.0 %
Melting point
195°C to 198°C
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text