SoftFit-L™ Pipette Tips, Filtered, Low-Retention, Hinged Rack, PCR Ready
Actual product may vary

SoftFit-L™ Pipette Tips, Filtered, Low-Retention, Hinged Rack, PCR Ready

Pipette samples with the accuracy and precision you expect from your pipette, every time, with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
2769-05-HR200 μL
2739-05-HR300 μL
2749-05-HR20 μL
2779-05-HR1000 μL
2789-05-HR1200 μL
Catalog number 2769-05-HR
HSN 84799090
Price (INR)
26,780.00
PK
Volume (Metric):
200 μL
Price (INR)
26,780.00
PK
Compatible with Rainin™ LTS pipettes. Provide outstanding accuracy and precision, every time.
  • Low-retention tips maximize sample delivery
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allows quick identification of tip volume
Specifications
Description200L, Filtered, Sterile, Hinged Rack
Volume (Metric)200 μL
FilteredFiltered
SterilitySterile
ColorNatural
Model200L
Length (English)1.97 in.
Length (Metric)5.00cm
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal
Color CodeGreen
For Use With (Equipment)Rainin LTS LiteTouch
Product LineART SoftFit L
Fit for PurposeSterile & PCR Ready
Unit SizePK