SoftFit-L™ Pipette Tips, Filtered, Low-Retention, Hinged Rack, PCR Ready
Thermo Scientific™

SoftFit-L™ Pipette Tips, Filtered, Low-Retention, Hinged Rack, PCR Ready

Pipette samples with the accuracy and precision you expect from your pipette, every time, with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
Have Questions?
變更視圖buttonViewtableView
產品號碼Volume (Metric)Unit Size
2769-05-HRPK200 μLPack of 960
2739-05-HR300 μLCase of 4800
2739-05-HRPK
亦稱為 2739-05-HR
300 μLPack of 960
2749-05-HR20 μLCase of 4800
2749-05-HRPK20 μLPack of 960
2769-05-HR200 μLCase of 4800
2779-05-HR1000 μLCase of 3072
2779-05-HRPK1000 μLPack of 768
2789-05-HR1200 μLCase of 3072
2789-05-HRPK
亦稱為 2789-05-HR
1200 μLPack of 768
產品號碼 2769-05-HRPK
價格 (HKD)
-
Volume (Metric):
200 μL
Unit Size:
Pack of 960
Compatible with Rainin™ LTS pipettes. Provide outstanding accuracy and precision, every time.
  • Low-retention tips maximize sample delivery
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allows quick identification of tip volume
規格
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description200L, Filtered, Sterile, Hinged Rack
Length (English)1.97 in.
Length (Metric)5.00cm
Model200L
ColorNatural
Color CodeGreen
FilteredFiltered
Fit for PurposeSterile & PCR Ready
For Use With (Equipment)Rainin LTS LiteTouch
FormatHinged Rack
Product LineART SoftFit L
SterilitySterile
Tip StyleUniversal
Volume (Metric)200 μL
Unit SizePack of 960