Quinoline Yellow, 95%, pure, water soluble
Quinoline Yellow, 95%, pure, water soluble
Quinoline Yellow, 95%, pure, water soluble
Thermo Scientific Chemicals

Quinoline Yellow, 95%, pure, water soluble

CAS: 8004-92-0 | C18H9NNa2O8S2 | 477.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 277340250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS8004-92-0
IUPAC Namedisodium 2-(1,3-dioxo-2,3-dihydro-1H-inden-2-yl)quinoline-6,8-disulfonate
Molecular FormulaC18H9NNa2O8S2
InChI KeyFZUOVNMHEAPVBW-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2N=C(C=CC2=C1)C1C(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=94 %
Appearance (Color)Yellow to orange-brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only