Thermo Scientific Chemicals

4-Carboxybenzo-18-crown-6, 99%, Thermo Scientific Chemicals

Catalog number: 277490010
1 g, Plastic bottle, Each
Thermo Scientific Chemicals

4-Carboxybenzo-18-crown-6, 99%, Thermo Scientific Chemicals

Catalog number: 277490010
1 g, Plastic bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
60835-75-8
IUPAC Name
2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine-18-carboxylate
Molecular Formula
C17H23O8
InChI Key
GFQYJVLOPVAPGJ-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=CC=C2OCCOCCOCCOCCOCCOC2=C1
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
Appearance (Form)
Powder
Melting point
121°C to 126°C
HPLC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text