2-Aminoterephthalic acid, 98.5%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 98.5%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 98.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Aminoterephthalic acid, 98.5%, Thermo Scientific Chemicals

CAS: 10312-55-7 | C8H5NO4 | 179.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 278031000
Price (USD)
234.65
Special offer
Online exclusive
Ends: 28-Dec-2026
276.00
Save 41.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
234.65
Special offer
Online exclusive
Ends: 28-Dec-2026
276.00
Save 41.35
Each
Chemical Identifiers
CAS10312-55-7
IUPAC Name2-aminobenzene-1,4-dicarboxylate
Molecular FormulaC8H5NO4
InChI KeyGPNNOCMCNFXRAO-UHFFFAOYSA-L
SMILESNC1=CC(=CC=C1C([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with NaOH98.0 to 102.0 %
HPLC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only