Petroleum ether, for analysis, boiling range 80-110°C
Petroleum ether, for analysis, boiling range 80-110°C
Petroleum ether, for analysis, boiling range 80-110°C
Thermo Scientific Chemicals

Petroleum ether, for analysis, boiling range 80-110°C

CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 278230010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS64742-49-0
IUPAC NamePetroleum ether
MDL NumberMFCD00081849
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Boiling point78°C to 113°C
Aromatics=<0.01 %
Cobalt (Co)=<0.1 ppm
Iron (Fe)=<0.2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only