Geranyl linalool, ca. 95%, Tech., mixture of isomers
Geranyl linalool, ca. 95%, Tech., mixture of isomers
Geranyl linalool, ca. 95%, Tech., mixture of isomers
Geranyl linalool, ca. 95%, Tech., mixture of isomers
Thermo Scientific Chemicals

Geranyl linalool, ca. 95%, Tech., mixture of isomers

CAS: 1113-21-9 | C20H34O | 290.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
25 g
Catalog number 278360250
Price (EUR)
654,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
654,00
Each
Add to cart
Chemical Identifiers
CAS1113-21-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Specific gravity(25°C) 0.875 to 0.885
Refractive index1.4860 to 1.4920 (20°C, 589 nm)
GC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only