Brucine, 99%, anhydrous
Brucine, 99%, anhydrous
Brucine, 99%, anhydrous
Thermo Scientific Chemicals

Brucine, 99%, anhydrous

CAS: 357-57-3 | C23H26N2O4 | 394.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 278721000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS357-57-3
IUPAC Name(1R,18S,20R,21R)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one
Molecular FormulaC23H26N2O4
InChI KeyRRKTZKIUPZVBMF-PLNGPGDESA-N
SMILESCOC1=CC2=C(C=C1OC)[C@@]13CCN4CC5=CCOC6CC(=O)N2C1[C@H]6[C@H]5C[C@@H]34
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.5 % (On dry substance)
Sulfated ash=<0.1 %
Specific optical rotation-118° to -128° (20°C, 589 nm) (c=2.5, CHCl3)
Appearance (Form)Crystalline powder
Loss on drying=<1 % (105°C, 3 hrs)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only