Octanophenone, 99+%
Octanophenone, 99+%
Octanophenone, 99+%
Acros Organics

Octanophenone, 99+%

CAS: 1674-37-9 | C14H20O | 204.31 g/mol
Have Questions?
Catalog NumberQuantityPackaging
279361000
also known as 27936
100 gGlass Bottle
Catalog number 279361000
also known as 27936
HSN 29143990
Price (INR)
39,839.00
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
39,839.00
Each
Chemical Identifiers
CAS1674-37-9
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5024 to 1.5054 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only