Silver sulfate, pure, 10 g/l solution in sulfuric acid
Silver sulfate, pure, 10 g/l solution in sulfuric acid
Silver sulfate, pure, 10 g/l solution in sulfuric acid
Thermo Scientific Chemicals

Silver sulfate, pure, 10 g/l solution in sulfuric acid

CAS: 10294-26-5 | Ag2H2O4S | 313.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number 279480025
Price (TWD)
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS10294-26-5
IUPAC Namedisilver(1+) sulfuric acid
Molecular FormulaAg2H2O4S
InChI KeyYPNVIBVEFVRZPJ-UHFFFAOYSA-N
SMILES[Ag+].[Ag+].OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Solution contains9.5 to 10.5 g/l Ag2SO4 in
Appearance (Color)Clear colorless
Solution containsH2SO4 95-97%
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only