Cyclohexane, 99.8%, for HPLC
Cyclohexane, 99.8%, for HPLC
Cyclohexane, 99.8%, for HPLC
Thermo Scientific Chemicals

Cyclohexane, 99.8%, for HPLC

Green features
CAS: 110-82-7, C6H12, 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 279590025
Price (USD)
235.00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
235.00
Each
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.8 %
Acidity (CH3COOH)=<0.0002 meq/g
UVat 230 nm A: =<0.02
UVat 250 nm A: =<0.005
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only