Search
View additional product information for RBS™ pF Detergent Concentrate - FAQs (27959, 27960)
1 product FAQs found
RBS-pF Detergent Concentrate (Cat. Nos. 27959 and 27960), and PCC-Pfree Detergent Concentrate (Cat. No. 72290) are all phosphate-free.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.