Ruthenium Red, ≥85%, pure
Ruthenium Red, ≥85%, pure
Ruthenium Red, ≥85%, pure
Thermo Scientific Chemicals

Ruthenium Red, ≥85%, pure

CAS: 11103-72-3 | H24Cl6N14O2Ru3 | 786.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 279710050
Price (EUR)
427,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
427,00
Each
Chemical Identifiers
CAS11103-72-3
IUPAC Nametris(λ²-ruthenium(2+)) tetradecaamine dihydrate hexachloride
Molecular FormulaCl6H46N14O2Ru3
InChI KeyJQJSTVUROJELSR-UHFFFAOYSA-H
SMILESN.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru++].[Ru++].[Ru++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Elemental Analysis34 to 39 % (Ru)
Appearance (Color)Dark red to brown to purple
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only