Thermo Scientific™
Richard-Allan Scientific™ Signature Series Clarifier™ 1, 2
Related applications:
Clinical
Enhance the nuclear definition and clarity of Signature Series hematoxylins with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Clarifier™ 1, 2.
Back to top
  • Improves contrast between Nuclei and Cytoplasm
  • Increases cell transparency
  • Removes background staining caused by excessive tissue adhesive
  • Removes unwanted hematoxylin staining from adhesive without affecting nuclear staining
Note:
  • Clarifier 1 is recommended with Hematoxylin 7211 and Hematoxylin 1
  • Clarifier 2 is recommended with Hematoxylin 7212 and Hematoxylin 2