POROS™ CaptureSelect™ AdV5 Affinity Matrix
Thermo Scientific™

POROS™ CaptureSelect™ AdV5 Affinity Matrix

Thermo Scientific™ POROS™ CaptureSelect™ AdV5 Affinity Matrix was designed specifically for the purification and/or scavenging of recombinant hexon capsomeres of Adenovirus serotype 5 (AdV5) particles, as well as free hexon proteins thereof, for high purity in a single step.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
280353201010 mL
28035320055 mL
280353205050 mL
Catalog number 2803532010
Price (USD)
2,400.00
Each
Quantity:
10 mL
Price (USD)
2,400.00
Each

POROS CaptureSelect AdV5 Affinity Matrix was designed specifically for the purification and/or scavenging of recombinant hexon capsomeres of Adenovirus serotype 5 (AdV5) particles, as well as free hexon proteins thereof, for high purity in a single step. This affinity resin specifically binds AdV5 virus particles and enables a robust, fast, and efficient purification for research applications with excellent purity and yield in one step.

Features of this affinity matrix include:

  • One-step purification of virus particles from crude material with high purity and yield
  • High specificity, capacity, and retained activity
  • Excellent scalability
  • Non-animal derived
  • High flow rates

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: cross-linked poly(styrene-divinylbenzene)
  • Average particle size: 50 μm
  • Ligand: POROS CaptureSelect AdV5 affinity ligand
  • Ligand coupling method: epoxide
  • Binding capacity: >1 x 1011 genome copies/mL of resin
  • Elution conditions: acidic elution with 0.1 M glycine, pH 3.0
  • Mechanical resistance: 100 bar (1450 psi, 10 MPa)
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Ligand TypeAdV5 affinity ligand
MatrixCross-linked poly(styrene-divinylbenzene)
Quantity10 mL
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Stationary PhaseAdV5
Binding Capacity>1 x 1011 Genome Copies/mL of Resin
FormResin Slurry
Particle Size50 μm
Product LineCaptureSelect, POROS
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light