PMPI (p-maleimidophenyl isocyanate)

Catalog number: 28100B

Thermo Scientific™  Related applications: Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce PMPI is a maleimide-and-isocyanate crosslinker for attaching compounds to sulfhydryl groups (cysteines) after conjugating the linker to hydroxyl groups on a molecule by urethane (carbamate) bonds.

Features of PMPI:

Reactive groups: maleimide and isocyanate
Reactive towards: sulfhydryl and hydroxyl groups
• Short (8.7A), sulfhydryl-to-hydroxyl crosslinker with an aromatic spacer arm (noncleavable)
• Maleimide group reacts with sulfhydryl groups to form stable thioether linkages
• Isocyanate reacts with non-aqueous hydroxyl groups to form carbamate linkage
• Excellent tool for conjugating hydroxyl-containing compounds such as steroids and vitamins
• Targets and reaction solutions must not contain primary amines
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Crosslinker Type: Heterobifunctional
Labeling Method: Chemical Labeling
Product Line: Pierce™
Product Size: Custom
Solubility: DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Spacer: Short

Contents & storage

Upon receipt store at 4°C protected from moisture. Product is shipped at ambient temperature.

Documents

Manuals & protocols