Hypersil™ BDS C8 HPLC Columns
Thermo Scientific™

Hypersil™ BDS C8 HPLC Columns

Use as a robust, general-purpose column for QC/QA applications. They are high-quality, base-deactivated, fully endcapped, have similar C18 selectivity but slightly less retentive.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
28203-05463050 mm4.6 mm3 μm
28205-05213050 mm2.1 mm5 μm
28205-05303050 mm3 mm5 μm
28203-07463075 mm4.6 mm3 μm
28205-05463050 mm4.6 mm5 μm
28205-102130100 mm2.1 mm5 μm
28203-103030100 mm3 mm3 μm
28205-104630100 mm4.6 mm5 μm
28203-104030100 mm4 mm3 μm
28205-154030150 mm4 mm5 μm
28205-124030125 mm4 mm5 μm
28205-154630150 mm4.6 mm5 μm
28205-124630125 mm4.6 mm5 μm
28205-254030250 mm4 mm5 μm
28205-254630250 mm4.6 mm5 μm
28205-01400110 mm4 mm5 μm
28203-154630150 mm4.6 mm3 μm
28203-153030150 mm3 mm3 μm
28203-104630100 mm4.6 mm3 μm
28203-05213050 mm2.1 mm3 μm
Catalog number 28203-054630
Price (HKD)
-
Length (Metric):
50 mm
Diameter (Metric):
4.6 mm
Particle Size:
3 μm

For a high-quality, base-deactivated, fully endcapped phase with similar selectivity to C18 but slightly less retention, use Thermo ScientificTM HypersilTM BDS C8 HPLC Columns. These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.

For more challenging separations that require improved chromatographic performance, use Thermo ScientificTM Hypersil GOLDTM C8 columns as an alternative.

  • Excellent choice for acids, bases and neutral compounds
  • Less retentive than C18 for faster analyses
  • Robust columns with long column lifetimes
  • Very reliable for routine assays

Ordering Information:

Other column dimensions are available. Please call Customer Service for more information.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeReversed Phase
EndcappedYes
For Use WithHPLC
Max. Pressure5800 psi (400 bar)
Packing MaterialSpherical, Fully Porous, Base-Deactivated Silica
Stationary PhaseC8
Surface Area170 m²/g
Temperature60°C
USP TypeL7
Carbon Load7%
Diameter (Metric)4.6 mm
Length (Metric)50 mm
Particle Size3 μm
PhaseReversed Phase
Pore Size130 Å
Product LineHypersil BDS
pH2 to 8
Unit SizeEach