Triton™ X-100 Surfact-Amps™ Detergent Solution
Thermo Scientific™

Triton™ X-100 Surfact-Amps™ Detergent Solution

Thermo Scientific Triton X-100 Surfact-Amps Detergent Solution is a highly-purified Triton™ X-100 detergent stabilized as a 10% solution (w/v) in variable sizes packaged under nitrogen in 10 mL glass ampules or three sizes of non-leaching HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
283131 L
8511150 mL
283146 x 10 mL
85112250 mL
카탈로그 번호 28313
제품 가격(KRW)
2,601,375
온라인 행사
Ends: 30-Sep-2026
2,973,000
할인액 371,625 (13%)
Each
수량:
1 L
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
2,601,375
온라인 행사
Ends: 30-Sep-2026
2,973,000
할인액 371,625 (13%)
Each

Thermo Scientific Triton X-100 Surfact-Amps Detergent Solution is a highly-purified Triton™ X-100 detergent stabilized as a 10% solution (w/v) in variable sizes packaged under nitrogen in 10 mL glass ampules or three sizes of non-leaching HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Features of Triton X-100 Surfact-Amps Detergent Solution:

  • Triton X-100—a nonionic detergent for use in various protein methods
  • Accurate—precise 10% detergent solution in ultrapure water
  • Easy-to-use—solution is simple to dispense and dilute for use
  • Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
  • Stable—packaged under inert nitrogen gas in glass ampules or HDPE bottles

Specifications for Triton X-100 Surfact-Amps Detergent Solution:

  • Visual: Clear to slightly hazy, colorless solution, free of particulate matter.
  • Concentration: 10.0±1.0%
  • Oxidants: ≤1.0μeq/mL
  • Carbonyls: ≤1.0μeq/mL
  • Suspended Solids: Residue present must not exceed Residue Reference.

Properties of Triton X-100:

  • Molecular Weight: 647 g
  • Detergent Class: Nonionic polyoxyethylene surfactant
  • Aggregation Number: 140
  • Micelle Molecular Weight: 90,000 g
  • Critical Micelle Concentration (CMC): 0.24 mM (0.0155% w/v)
  • Cloud Point: 64°C
  • Dialyzable: No

Triton X-100 is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins.

Related Products

  • Surfact-Amps™ Detergent Sampler (Cat. No. 28340)
For Research Use Only. Not for use in diagnostic procedures.

Specifications:

  • Visual: Clear to slightly hazy, colorless solution, free of particulate matter.
  • Concentration: 10.0 ± 1.0%
  • Oxidants: ≤ 1.0 μeq/mL
  • Carbonyls: ≤ 1.0 μeq/mL
  • Suspended Solids: Residue present must not exceed Residue Reference.
사양
수량1 L
시약 유형Detergent Solution
형태Liquid
제품라인Surfact-Amps, Triton
Unit SizeEach
구성 및 보관
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.

자주 묻는 질문(FAQ)

What are the storage conditions for Triton X-100 Surfact-Amps Detergent Solution (Cat. No. 85111)?

After opening, we recommend storing the unused solution sealed in an air-tight vial containing minimal air space or from which the air has been purged with nitrogen gas. Frequent opening should be avoided because Triton X-100 is susceptible to autoxidation upon exposure to air.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can you provide the shelf life for Triton X-100 Surfact-Amps Detergent Solution (Cat. No. 85111)?

Triton X-100 Surfact-Amps Detergent Solution is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (20-25 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.