Triton™ X-100 Surfact-Amps™ Detergent Solution

Catalog number:  28314B

Thermo Scientific™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Triton X-100 Surfact-Amps Detergent Solution is a highly-purified Triton™ X-100 detergent stabilized as a 10% solution in variable sizes packaged under nitrogen in 10 mL glass ampules or three sizes of non-leaching HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Features of Triton X-100 Surfact-Amps Detergent Solution:

Triton X-100—a nonionic detergent for use in various protein methods
Accurate—precise 10% detergent solution in ultrapure water
Easy-to-use—solution is simple to dispense and dilute for use
Exceptionally pure—less than 1.0µeq/mL peroxides and carbonyls
Stable—packaged under inert nitrogen gas in glass ampules or HDPE bottles

Specifications for Triton X-100 Surfact-Amps Detergent Solution:
• Visual: Clear to slightly hazy, colorless solution, free of particulate matter.
• Concentration: 10.0±1.0%
• Oxidants: ≤1.0µeq/mL
• Carbonyls: ≤1.0µeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference.

Properties of Triton X-100:
• Molecular Weight: 647g
• Detergent Class: Nonionic polyoxyethylene surfactant
• Aggregation Number: 140
• Micelle Molecular Weight: 90,000g
• Critical Micelle Concentration (CMC): 0.24 mM (0.155%, w/v)
• Cloud Point: 64°C
• Dialyzable: No

Triton X-100 is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins.

Related Products
Surfact-Amps™ Detergent Sampler
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Line: Surfact-Amps™, Triton™
Product Size: Custom
Reagent Type: Detergent Solution

Contents & storage

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.

Documents

Manuals & protocols