Brij™-35 Surfact-Amps™ Detergent Solution

Catalog number: 28316B

Thermo Scientific™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Brij™ 35 Surfact-Amps™ Detergent Solution is a stabilized 10% solution of Brij-35 detergent for use in various protein methods. Thermo Scientific™ Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability.

Properties of Brij 35 Detergent:

• Molecular Weight: 1225g
• Detergent Class: Nonionic
• Aggregation Number: 40
• Micelle Molecular Weight: 49,000g
• Critical Micelle Concentration (CMC): 0.09 mM (0.011%, w/v)
• Cloud Point: >100°C
• Dialyzable: No

The detergent Brij™ 35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or a surfactant in various HPLC applications. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules or HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Features of these Brij 35 Surfact-Amps Detergent Solutions:

Brij 35—a nonionic detergent for use in various protein methods
Accurate—10% detergent solutions in ultrapure water
Easy-to-use—solutions are simple to dispense and dilute for use

Specifications Brij 35 Surfact-Amps Detergent Solutions:

• Visual: Clear, colorless solution, free of particulate matter.
• Concentration: 10.0 ±1.0%
• Oxidants: ≤1.0µeq/mL
• Carbonyls: ≤1.0µeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference

Related Products
Brij™-35, 30% Solution

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Line: Brij™, Surfact-Amps™
Product Size: Custom
Reagent Type: Detergent Solution

Contents & storage

Store at room temperature.

Documents

Manuals & protocols