NP-40 Surfact-Amps™ Detergent Solution

Catalog number: 28324B

Thermo Scientific™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized as a 10% solution in your choice of either 10 mL glass ampules or two sizes of HDPE-plastic bottles.

Features of Surfact-Amps NP-40 Detergent Solution:

NP-40—a nonionic detergent for use in various protein methods
Accurate—precise 10% detergent solutions in ultrapure water
Easy-to-use—solutions are simple to dispense and dilute for use
Exceptionally pure—less than 1.0µeq/mL peroxides and carbonyls
Stable—packaged under inert nitrogen gas in HDPE bottles

This is an easy-to-use 10% (w/v) solution of purified NP-40 detergent for use in routine and high-demand protein research methods and molecular biology techniques. NP-40 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins. Surfact-Amps Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules or non-leaching HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Properties of NP-40 Detergent:
• Molecular Weight: 617g
• Detergent Class: Nonionic
• Aggregation Number: 149
• Micelle Molecular Weight: 90,000g
• Critical Micelle Concentration (CMC): 0.29 mM (0.0179%, w/v)
• Cloud Point: 80°C
• Dialyzable: No

Specifications for NP-40 Surfact-Amps Detergent Solution:
• Visual: Clear, colorless solution, free of particulate matter.
• Concentration: 10.0±1.0%
• Oxidants: ≤1.0µeq/mL
• Carbonyls: ≤1.0µeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference.

Related Products
Surfact-Amps™ Detergent Sampler
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Line: Surfact-Amps™
Product Size: Custom
Reagent Type: Detergent Solution

Contents & storage

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.

Documents

Manuals & protocols