Limited time offer: 3 year warranty when you purchase a SimpliAmp Thermal Cycler. View flyer »

Tween™ 80 Surfact-Amps™ Detergent Solution

Catalog number: 28328B

Thermo Scientific™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Tween-80 Surfact-Amps Detergent Solution is a highly-purified Tween™ 80 detergent stabilized as a 10% solution and packaged under nitrogen in glass ampules or non-leaching HDPE bottles, ensuring it's stability and eliminating the accumulation of peroxides and degradation products.

Features of Tween-80 Surfact-Amps Detergent Solution:

Tween 80—a popular nonionic detergent for use in ELISA, Western blotting and other immunoassay PBS or TBS wash buffers
Accurate—precise 10% detergent solution in ultrapure water
Easy-to-use—solution is simple to dispense and dilute for use
Exceptionally pure—less than 1.0µeq/mL peroxides and carbonyls
Stable—packaged under inert nitrogen gas in glass ampules or HDPE bottles

Properties of Tween 80:
• Molecular Weight: 1310g
• Detergent Class: Nonionic polyoxyethylene surfactant
• Aggregation Number: 60
• Micelle Molecular Weight: 76,000g
• Critical Micelle Concentration (CMC): 0.012 mM (0.0016%, w/v)
• Cloud Point: Unknown
• Dialyzable: No

Specifications for Tween 80 Surfact-Amps Detergent Solution:
• Visual: Slightly hazy to cloudy, colorless solution, free of particulates; may form layers.
• Concentration: 10.0±1.0%
• Oxidants: ≤1.0µeq/mL
• Carbonyls: ≤1.0µeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference.

Related Products
Surfact-Amps™ Detergent Sampler
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Line: Surfact-Amps™, Tween™
Product Size: Custom
Reagent Type: Detergent Solution

Contents & storage

Store in a cool, dry, and well-ventilated area

Documents

Manuals & protocols