Pierce™ 10X Tris-Glycine Buffer, 1 L - FAQs
View additional product information for Pierce™ Concentrated Buffer Stocks (10X and 20X) - FAQs (28352, 28341, 28363, 28362, 28346, 28354, 28344, 28355, 28358, 28348, 28360)
1 product FAQs found
What is the optimal pH for the Pierce Concentrated Buffer Stocks (10X and 20X) (Cat. No. 28362)?
The quality specifications for the ready to use 1x solution is 8.5 ± 0.2. (see example COA: https://assets.thermofisher.com/TFS-Assets/LSG/certificate/Certificates-of-Analysis/28362_WH332690.PDF). The pH should be between 8.3 and 8.7.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.