Search
View additional product information for Dry-blend Buffered Packs - FAQs (28374, 28372, 28384, 28379, 28376, 28382, 28390, 28380)
1 product FAQs found
The BupH Borate Buffered Saline Packs is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.