Precision™ Concentric Ring Electrical Steaming Baths
El producto real puede variar
Thermo Scientific™

Precision™ Concentric Ring Electrical Steaming Baths

Thermo Scientific™ Precision™ concentric ring electrical steaming water baths have been discontinued.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)N.º de posicionesRequisitos eléctricos
289915 L8230 V, 50/60 Hz
2896TS
también denominado 2896
7,5 L4120 V 50/60 Hz
28977,5 L4230 V, 50/60 Hz
289815 L8120 V 50/60 Hz
Número de catálogo 2899
Precio (MXN)
-
Capacidad (métrico):
15 L
N.º de posiciones:
8
Requisitos eléctricos:
230 V, 50/60 Hz

The Thermo Scientific™ Precision™ concentric ring electrical steaming water baths have been discontinued. For your water bath needs, please see our current offerings of Thermo Scientific water baths.

Baths accommodate various sizes of glassware and evaporating dishes, including round-bottom flasks. Flange-formed stainless-steel rings nest smoothly and become progressively larger as each concentric ring is removed. Copper-clad immersion heaters provide bath temperatures up to 100°C.

  • 4- and 8-hole models enlarge from 0.88in. (2.2cm) to maximum opening diameters of 5 and 6in. (12.7 and 15.2cm), respectively
  • Copper-clad immersion heaters provide bath temperature up to 100°C
  • Center ring cover is 1in.(2.5cm) in diameter
  • Easy to operate with a four-position switch and three different heat settings
  • Adjustable water level regulator maintains bath depth at the desired level
  • Stainless-steel cabinets with flanged cabinet top for tabletop installations
  • Rugged epoxy-coated, corrosion-resistant stand
  • Accommodate most laboratory glassware
  • Baths may also be flush-mounted in benchtop

Ordering Information:

Wiring of switch must be completed onsite.

Includes:

Precision ring sets, constant water level device (with 0.5in. inlet for water supply), adjustable drain tube, and welded angle-iron support frame with hot-rolled steel legs, fasteners and lockwashers

Requires:

Hardwire installation by a qualified electrician and hook-up to a continuous water source

Recommended for:

  • Copper strip corrosion tests
  • Melting agar
  • Metallurgical analysis
  • Thawing cryopreservation vials
  • Warranty varies by country, please contact us for details
Especificaciones
Amperaje8.3 A
Diámetro de la apertura mín. (imperial)0.88 in.
Diámetro de la apertura mín. (métrico)122 mm
Frecuencia50/60 Hz
Tiempo de calentamiento65 min. to 100°C
Ajustes del calentador (bajo, medio, alto)500w, 1000w, 2000w (Low, Medium, High)
Altura exterior con soporte (imperial)13.5 in.
Altura exterior sin soporte (imperial)4.5 in.
Altura exterior con soporte (métrico)34.3 cm
Altura exterior sin soporte (métrico)11.4 cm
Longitud exterior con soporte (imperial)15.5 in.
Longitud exterior sin soporte (imperial)15.5 in.
Longitud exterior con soporte (métrico)39.4 cm
Longitud exterior sin soporte (métrico)39.4 cm
MaterialAcero inoxidable
Diámetro mín. de la abertura (sin ninguna anilla)2 at 100 mm (4 in.); 4 at 125 mm (5 in.); 2 at 150 mm (6 in.)
N.º de posiciones8
Temperatura máxima (métrico)100 °C
Tensión230 V
Anchura exterior con soporte (imperial)19 in.
Anchura exterior sin soporte (imperial)29 in.
Anchura exterior con soporte (métrico)73.7 cm
Anchura exterior sin soporte (métrico)73.7 cm
Capacidad (métrico)15 L
Requisitos eléctricos230 V, 50/60 Hz
Unit SizeEach