規格
Certifications/ComplianceIncludes Certificate of Analysis
DescriptionPower-Bind Streptavidin Coated Particles
IncludesPackage Insert and Certificate of Analysis
Preservative0.05% Na Az
Quantity1 mL
Sample Evaluation PacksPower-Bind evaluation samples are packaged in 1mL bottles at 1% solids (10 mg/mL). Select individual lots to evaluate.
Shelf Life24 Months
Specialty PackagingPower-Bind Streptavidin Coated particles are available in different concentrations and package sizes upon request.
StabilityGuanidinium thiocyanate; DMF and DMSO to 100% Sonication; environments PCR temperature cycling
ColorWhite or Colorless
Diameter (Metric)0.3 μm
For Use With (Application)Biotinylated Ligand Binding, Diagnostic Assays
MaterialPolystyrene
Product TypeStreptavidin-Coated Particles
Unit SizeEach