GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Actual product may vary
Thermo Scientific™
GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
29001320TSLiner Sealing RingTRACE 1300 Series GC SSL Injectors
29033406Liner Sealing RingTRACE and FOCUS GC SSL Injectors
290GA243Graphite Liner Sealing RingAgilent SSL Liners
290OA241Viton Liner Sealing RingTRACE 1300 Series GC and Agilent SSL Injectors
29001318TS
also known as 29001318
Liner Sealing RingTRACE 1300 Series GC PTV Injectors
Catalog number 29001320TS
Price (EUR)
-
Description:
Liner Sealing Ring
For Use With (Equipment):
TRACE 1300 Series GC SSL Injectors
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
  • These liner sealing rings are designed for optimal performance in gas chromatography systems.
    • Product Type: Liner Sealing Ring
    • For Use With: TRACE 1300 Series GC PTV Injectors
Specifications
DescriptionLiner Sealing Ring
For Use With (Equipment)TRACE 1300 Series GC SSL Injectors
Quantity5 Pack
Product TypeLiner Sealing Ring
Unit SizePack of 5