L(+)-Isovaline monohydrate, 99+%, (>99% ee)
L(+)-Isovaline monohydrate, 99+%, (>99% ee)
L(+)-Isovaline monohydrate, 99+%, (>99% ee)
Thermo Scientific Chemicals

L(+)-Isovaline monohydrate, 99+%, (>99% ee)

CAS: 595-40-4 | C5H11NO2 | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 290052500
Price (USD)
261.65
Special offer
Online exclusive
Ends: 28-Dec-2026
308.00
Save 46.35
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
261.65
Special offer
Online exclusive
Ends: 28-Dec-2026
308.00
Save 46.35
Each
Chemical Identifiers
CAS1266371-20-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO499.0 to 101.0 %
HPLC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only