100% Graphite Ferrules for Thermo Scientific Instruments
Thermo Scientific™
100% Graphite Ferrules for Thermo Scientific Instruments
Ensure reliable connections for Thermo Scientific™ GC instruments with these Thermo Scientific™ 100% Graphite Ferrules for Thermo Scientific Instruments.
Catalog NumberFor Use With (Equipment)Diameter (Metric)
290GA108Agilent Injectors and Non-MS Detectors3.175 mm
29053487Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors0.32 mm
29053488Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors0.1 to 0.25 mm
290GA107Agilent Injectors and Non-MS Detectors6.35 mm
Catalog number 290GA108
Price (GBP)
-
For Use With (Equipment):
Agilent Injectors and Non-MS Detectors
Diameter (Metric):
3.175 mm
Ensure optimal performance of your Thermo Scientific™ GC instrument with Thermo Scientific™ Ferrules. They are available in three different materials and in various dimensions to accommodate a range of instruments and applications. All ferrules are are supplied in contaminant-free, individual blister packs.
Features
  • Available in 100% graphite, 15% graphite/85% vespel and SilTite metal
  • Ferrules for different types of injectors and detectors
Specifications
Description1/8 in. O.D.; Agilent 5890, 6890, 6850
Diameter (English)1/8 in.
Diameter (Metric)3.175 mm
For Use With (Equipment)Agilent Injectors and Non-MS Detectors
Material100% Graphite
TypePacked Column
Unit SizePack of 10