GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Thermo Scientific™
GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
290OA241Viton Liner Sealing RingTRACE 1300 Series GC and Agilent SSL Injectors
29033406Liner Sealing RingTRACE and FOCUS GC SSL Injectors
290GA243Graphite Liner Sealing RingAgilent SSL Liners
29001318TS
also known as 29001318
Liner Sealing RingTRACE 1300 Series GC PTV Injectors
29001320TSLiner Sealing RingTRACE 1300 Series GC SSL Injectors
Catalog number 290OA241
Price (USD)
40.37
Pack of 10
Description:
Viton Liner Sealing Ring
For Use With (Equipment):
TRACE 1300 Series GC and Agilent SSL Injectors
Price (USD)
40.37
Pack of 10
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Specifications
DescriptionViton Liner Sealing Ring
For Use With (Equipment)TRACE 1300 Series GC and Agilent SSL Injectors
Quantity10 Pack
Product TypeViton Liner Sealing Ring
Unit SizePack of 10