Tris(trimethylsilyl)silane, 96%
Tris(trimethylsilyl)silane, 96%
Tris(trimethylsilyl)silane, 96%
Thermo Scientific Chemicals

Tris(trimethylsilyl)silane, 96%

CAS: 1873-77-4 | C9H28Si4 | 248.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 mL
25 mL
Packaging:
Glass bottle
Catalog number 291060250
Price (TWD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1873-77-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4890 to 1.4910 (20°C, 589 nm)
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only