Octil 4-metoxicinamato, 98 %, estabilizado, Thermo Scientific Chemicals
Octil 4-metoxicinamato, 98 %, estabilizado, Thermo Scientific Chemicals
Octil 4-metoxicinamato, 98 %, estabilizado, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Octil 4-metoxicinamato, 98 %, estabilizado, Thermo Scientific Chemicals

CAS: 5466-77-3 | C18H26O3 | 290.40 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
Envase:
Frasco de vidrio
Número de catálogo 291161000
Precio (EUR)
188,00
Each
Cantidad:
100 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
188,00
Each
Identificadores químicos
CAS5466-77-3
IUPAC Name2-ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate
Molecular FormulaC18H26O3
InChI KeyYBGZDTIWKVFICR-UHFFFAOYNA-N
SMILESCCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Liquid
Specific gravity(25/25°C) 1.007 to 1.012
Stabilizer0.05 to 0.1 % BHT
Appearance (Color)Clear colorless to yellow
GC>=97.5 %
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only